2-Desoxy-D-glucose
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
(3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal
AS35020G1
AS35020G10
AS35020G5
ACTIAS35020G1EA
0
CHF
ACTIAS35020G1
ACTIAS35020G10
ACTIAS35020G5
2-Desoxy-D-glucose
2-Deoxy-D-glucose
Formel:
C₆H₁₂O₅ MW: 164.16 g/mol |
MDL Number:
MFCD00151328 CAS-Nummer: 154-17-6 EINECS: 205-823-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...