3-Amino-5-brom-2-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-2-fluoropyridin-3-amine

AS33448G1 AS33448G10 AS33448G5
ACTIAS33448G1EA 0 CHF
ACTIAS33448G1 ACTIAS33448G10 ACTIAS33448G5
3-Amino-5-brom-2-fluorpyridin
3-Amino-5-bromo-2-fluoropyridine
Formel: C₅H₄BrFN₂
MW: 191.0011 g/mol
MDL Number: MFCD06659524
CAS-Nummer: 884495-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR