2-Brom-4-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS50957G1 AS50957G5
ACTIAS50957G1EA 21 CHF
ACTIAS50957G1 ACTIAS50957G5
2-Brom-4-fluorpyridin
2-Bromo-4-fluoropyridine
Formel: C₅H₃BrFN
MW: 175,99 g/mol
MDL Number: MFCD09260909
CAS-Nummer: 357927-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR