2,5-Dibrom-3-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS51020G1 AS51020G5
ACTIAS51020G1EA 0 CHF
ACTIAS51020G1 ACTIAS51020G5
2,5-Dibrom-3-fluorpyridin
2,5-Dibromo-3-fluoropyridine
Formel: C₅H₂Br₂FN
MW: 254,88 g/mol
MDL Number: MFCD06659497
CAS-Nummer: 156772-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR