3-Fluor-5-methoxybenzylbromid

Lieferant: ACTIVATE SCIENTIFIC

AS32630G1 AS32630G5
ACTIAS32630G1EA 0 CHF
ACTIAS32630G1 ACTIAS32630G5
3-Fluor-5-methoxybenzylbromid
3-Fluor-5-methoxybenzylbromid
Formel: C₈H₈BrFO
MDL Number: MFCD08741378
CAS-Nummer: 914637-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR