4-(Tributylstannyl)pyridin
Lieferant: ACTIVATE SCIENTIFIC
|
Gefahr
|
Synonyms:
4-(Tri-n-butylstannyl)pyridine
AS50170G1
AS50170G25
AS50170G5
ACTIAS50170G1EA
72
CHF
ACTIAS50170G1
ACTIAS50170G25
ACTIAS50170G5
4-(Tributylstannyl)pyridin
4-(Tributylstannyl)pyridin
Formel:
C₁₇H₃₁NSn MW: 368,15 g/mol |
MDL Number:
MFCD01319071 CAS-Nummer: 124252-41-1 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...