2-Brom-6-(tributylstannyl)pyridin

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51481G1 AS51481G5
ACTIAS51481G1EA 0 CHF
ACTIAS51481G1 ACTIAS51481G5
2-Brom-6-(tributylstannyl)pyridin
2-Brom-6-(tributylstannyl)pyridin
Formel: C₁₇H₃₀BrNSn
CAS-Nummer: 189083-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR