4-(2-Thienyl)buttersäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(Thiophen-2-yl)butanoic acid

AS22125G25 AS22125G5
ACTIAS22125G25EA 0 CHF
ACTIAS22125G25 ACTIAS22125G5
4-(2-Thienyl)buttersäure
4-(2-Thienyl)buttersäure
Formel: C₈H₁₀O₂S
MW: 170,23 g/mol
MDL Number: MFCD00005463
CAS-Nummer: 4653-11-6
EINECS: 225-090-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR