(R)-(+)-Indolin-2-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS20998G25 AS20998G5
ACTIAS20998G25EA 942 CHF
ACTIAS20998G25 ACTIAS20998G5
(R)-(+)-Indolin-2-carbonsäure
(R)-(+)-Indolin-2-carbonsäure
Formel: C₉H₉NO₂
MW: 163,18 g/mol
MDL Number: MFCD00792496
CAS-Nummer: 98167-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR