5-Chlor-3-methylbenzo[b]thiophen

Lieferant: ACTIVATE SCIENTIFIC

AS35028G1 AS35028G10 AS35028G5
ACTIAS35028G1EA 0 CHF
ACTIAS35028G1 ACTIAS35028G10 ACTIAS35028G5
5-Chlor-3-methylbenzo[b]thiophen
5-Chlor-3-methylbenzo[b]thiophen
Formel: C₉H₇ClS
MW: 182.67 g/mol
MDL Number: MFCD00052506
CAS-Nummer: 19404-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR