Methyl-3-amino-2-thiophencarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 3-amino-2-thiophenecarboxylate

AS2076G100 AS2076G25
ACTIAS2076G100EA 0 CHF
ACTIAS2076G100 ACTIAS2076G25
Methyl-3-amino-2-thiophencarboxylat
Methyl-3-amino-2-thiophencarboxylat
Formel: C₆H₇NO₂S
MW: 157,19 g/mol
MDL Number: MFCD00009765
CAS-Nummer: 22288-78-4
EINECS: 244-895-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR