6-Methoxyisochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS1986G1 AS1986G5
ACTIAS1986G1EA 0 CHF
ACTIAS1986G1 ACTIAS1986G5
6-Methoxyisochinolin
6-Methoxyisoquinoline
Formel: C₁₀H₉NO
MW: 159,1846 g/mol
MDL Number: MFCD04114861
CAS-Nummer: 52986-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR