2-(Methoxycarbonyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methoxycarbonylphenylboronic acid, 2-(Methoxycarbonyl)benzeneboronic acid

AS50858G1 AS50858G100 AS50858G5
ACTIAS50858G1EA 0 CHF
ACTIAS50858G1 ACTIAS50858G100 ACTIAS50858G5
2-(Methoxycarbonyl)phenylboronsäure
2-(Methoxycarbonyl)phenylboronsäure
Formel: C₈H₉BO₄
MW: 179,97 g/mol
MDL Number: MFCD02179452
CAS-Nummer: 374538-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR