Glycinbenzylester Hydrochlorid (H-Gly-OBzl.HCl)
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
H-Gly-OBzl·HCl, H-DL-Gly-OBzl hydrochloride, Benzyl glycinate hydrochloride, H-DL-Gly-OBzl.HCl
AS34252G10
AS34252G100
AS34252G5
ACTIAS34252G10EA
0
CHF
ACTIAS34252G10
ACTIAS34252G100
ACTIAS34252G5
Glycinbenzylester Hydrochlorid (H-Gly-OBzl.HCl)
Glycinbenzylester Hydrochlorid (H-Gly-OBzl.HCl)
Formel:
C₉H₁₂ClNO₂ MW: 201,65 g/mol |
MDL Number:
MFCD00035442 CAS-Nummer: 2462-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...