Triethylmethantricarboxylat

Lieferant: ACTIVATE SCIENTIFIC

AS25115G100 AS25115G25
ACTIAS25115G100EA 0 CHF
ACTIAS25115G100 ACTIAS25115G25
Triethylmethantricarboxylat
Triethylmethantricarboxylat
Formel: C₁₀H₁₆O₆
MDL Number: MFCD00009150
CAS-Nummer: 6279-86-3
EINECS: 228-477-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR