3-Methyl-3-oxetanmethanol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Methyl-3-oxethanemethanol

AS21209G100 AS21209G25
ACTIAS21209G100EA 66 CHF
ACTIAS21209G100 ACTIAS21209G25
3-Methyl-3-oxetanmethanol
3-Methyl-3-oxetanmethanol
Formel: C₅H₁₀O₂
MW: 102,13 g/mol
MDL Number: MFCD00010273
CAS-Nummer: 3143-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR