7-Bromisochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS1434G1 AS1434G5
ACTIAS1434G1EA 0 CHF
ACTIAS1434G1 ACTIAS1434G5
7-Bromisochinolin
7-Bromoisoquinoline
Formel: C₉H₆BrN
MW: 208,0600 g/mol
MDL Number: MFCD07368661
CAS-Nummer: 58794-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR