6-Bromisochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS1286G1 AS1286G5
ACTIAS1286G1EA 0 CHF
ACTIAS1286G1 ACTIAS1286G5
6-Bromisochinolin
6-Bromoisoquinoline
Formel: C₉H₆BrN
MDL Number: MFCD04973299
CAS-Nummer: 34784-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR