4-Chlor-3-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-Chloro-3-methoxyphenyl)boronic acid

AS33917G1 AS33917G5
ACTIAS33917G1EA 0 CHF
ACTIAS33917G1 ACTIAS33917G5
4-Chlor-3-methoxyphenylboronsäure
4-Chlor-3-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
MW: 186,4000 g/mol
MDL Number: MFCD01318965
CAS-Nummer: 89694-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR