5-Chlor-2-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (5-Chloro-2-methoxyphenyl)boronic acid, 5-Chloro-2-methoxybenzeneboronic acid

AS24346G100 AS24346G25
ACTIAS24346G100EA 191 CHF
ACTIAS24346G100 ACTIAS24346G25
5-Chlor-2-methoxyphenylboronsäure
5-Chlor-2-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
MW: 186.40 g/mol
MDL Number: MFCD01318966
CAS-Nummer: 89694-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR