2-Chlor-4-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-4-methoxybenzeneboronic acid

AS31536G1 AS31536G5
ACTIAS31536G1EA 0 CHF
ACTIAS31536G1 ACTIAS31536G5
2-Chlor-4-methoxyphenylboronsäure
2-Chlor-4-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
MW: 186,40 g/mol
MDL Number: MFCD03411935
CAS-Nummer: 219735-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR