4-Iodbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS34890G1 AS34890G10 AS34890G5
ACTIAS34890G1EA 0 CHF
ACTIAS34890G1 ACTIAS34890G10 ACTIAS34890G5
4-Iodbenzaldehyd
4-Iodobenzaldehyde
Formel: C₇H₅IO
MW: 232.02 g/mol
MDL Number: MFCD00039576
CAS-Nummer: 15164-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR