1-(4-Nitrophenyl)pyrrolidin

Lieferant: ACTIVATE SCIENTIFIC

AS30312G1 AS30312G5
ACTIAS30312G1EA 0 CHF
ACTIAS30312G1 ACTIAS30312G5
1-(4-Nitrophenyl)pyrrolidin
1-(4-Nitrophenyl)pyrrolidin
Formel: C₁₀H₁₂N₂O₂
MW: 192,214 g/mol
MDL Number: MFCD00020819
CAS-Nummer: 10220-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR