2,5-Dibromnicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

AS32339G1 AS32339G5
ACTIAS32339G1EA 0 CHF
ACTIAS32339G1 ACTIAS32339G5
2,5-Dibromnicotinsäure
2,5-Dibromnicotinsäure
Formel: C₆H₃Br₂NO₂
MW: 280.9000 g/mol
MDL Number: MFCD06254512
CAS-Nummer: 29312-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR