Ethinyltributylzinn

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS50168G1 AS50168G5
ACTIAS50168G1EA 0 CHF
ACTIAS50168G1 ACTIAS50168G5
Ethinyltributylzinn
Ethynyltributyltin
Formel: C₁₄H₂₈Sn
MDL Number: MFCD00009420
CAS-Nummer: 994-89-8
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR