4-Fluor-3-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-Fluoro-3-methoxyphenyl)boronic acid, 4-Fluoro-3-methoxybenzeneboronic acid

AS32644G1 AS32644G5
ACTIAS32644G1EA 0 CHF
ACTIAS32644G1 ACTIAS32644G5
4-Fluor-3-methoxyphenylboronsäure
4-Fluor-3-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
MDL Number: MFCD08056358
CAS-Nummer: 854778-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR