4-Fluor-2-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Fluoro-2-methoxybenzeneboronic acid

AS51385G1 AS51385G5
ACTIAS51385G1EA 0 CHF
ACTIAS51385G1 ACTIAS51385G5
4-Fluor-2-methoxyphenylboronsäure
4-Fluor-2-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
MW: 169,95 g/mol
MDL Number: MFCD03788423
CAS-Nummer: 179899-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR