3-Fluor-5-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Fluoro-5-methoxybenzeneboronic acid

AS51162G1 AS51162G25 AS51162G5
ACTIAS51162G1EA 31 CHF
ACTIAS51162G1 ACTIAS51162G25 ACTIAS51162G5
3-Fluor-5-methoxyphenylboronsäure
3-Fluor-5-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
MW: 169,95 g/mol
MDL Number: MFCD07363780
CAS-Nummer: 609807-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR