2-Fluor-6-methoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-6-methoxybenzeneboronic acid

AS51375G100 AS51375G5
ACTIAS51375G100EA 0 CHF
ACTIAS51375G100 ACTIAS51375G5
2-Fluor-6-methoxyphenylboronsäure
2-Fluor-6-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
MW: 169,95 g/mol
MDL Number: MFCD02179483
CAS-Nummer: 78495-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR