3-(3,5-Dimethoxyphenyl)propionsäure

Lieferant: ACTIVATE SCIENTIFIC

AS33284G1 AS33284G5
ACTIAS33284G1EA 0 CHF
ACTIAS33284G1 ACTIAS33284G5
3-(3,5-Dimethoxyphenyl)propionsäure
3-(3,5-Dimethoxyphenyl)propionsäure
Formel: C₁₁H₁₄O₄
MW: 210,23 g/mol
MDL Number: MFCD03425685
CAS-Nummer: 717-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR