(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
(S)-1,2,3,4-Tetrahydronaphthalen-1-amine
AS32324G1
AS32324G10
AS32324G5
ACTIAS32324G1EA
0
CHF
ACTIAS32324G1
ACTIAS32324G10
ACTIAS32324G5
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamin
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamin
Formel:
C₁₀H₁₃N MW: 147,22 g/mol |
MDL Number:
MFCD00671630 CAS-Nummer: 23357-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...