4-Hydroxy-2(5H)-furanon (Tetrahydrofuran-2,4-dion, Tetronsäure, β-Oxo-γ-butyrolacton)

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Tetronic acid, beta-Tetronic acid, 2,4(3H,5H)-Furandione, Tetrahydrofuran-2,4-dione

AS35827G1 AS35827G5
ACTIAS35827G1EA 0 CHF
ACTIAS35827G1 ACTIAS35827G5
4-Hydroxy-2(5H)-furanon (Tetrahydrofuran-2,4-dion, Tetronsäure, β-Oxo-γ-butyrolacton)
4-Hydroxy-2(5H)-furanon (Tetrahydrofuran-2,4-dion, Tetronsäure, β-Oxo-γ-butyrolacton)
Formel: C₄H₄O₃
MW: 100.07 g/mol
MDL Number: MFCD00052187
CAS-Nummer: 541-57-1
EINECS: 208-785-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR