2-Chlor-N,N-dimethylacetamid
Lieferant: ACTIVATE SCIENTIFIC
AS21223G25
AS21223G5
ACTIAS21223G25EA
40
CHF
ACTIAS21223G25
ACTIAS21223G5
2-Chlor-N,N-dimethylacetamid
2-Chlor-N,N-dimethylacetamid
Formel:
C₄H₈ClNO MW: 121,57 g/mol |
MDL Number:
MFCD00039360 CAS-Nummer: 2675-89-0 EINECS: 220-224-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...