2-Brom-4-methylpyrimidin

Lieferant: ACTIVATE SCIENTIFIC

AS31050G1 AS31050G5
ACTIAS31050G1EA 0 CHF
ACTIAS31050G1 ACTIAS31050G5
2-Brom-4-methylpyrimidin
2-Bromo-4-methylpyrimidine
Formel: C₅H₅ClN₂
MDL Number: MFCD04037883
CAS-Nummer: 130645-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR