2-Pyrimidincarbonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Cyanopyrimidine

AS34178G10 AS34178G100
ACTIAS34178G10EA 0 CHF
ACTIAS34178G10 ACTIAS34178G100
2-Pyrimidincarbonitril
2-Pyrimidincarbonitril
Formel: C₅H₃N₃
MW: 105,1 g/mol
MDL Number: MFCD00160513
CAS-Nummer: 14080-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR