2-Chlor-5-methoxypyrimidin

Lieferant: ACTIVATE SCIENTIFIC

AS31800G1 AS31800G5
ACTIAS31800G1EA 0 CHF
ACTIAS31800G1 ACTIAS31800G5
2-Chlor-5-methoxypyrimidin
2-Chloro-5-methoxypyrimidine
Formel: C₅H₅ClN₂O
MW: 144,56 g/mol
MDL Number: MFCD08702770
CAS-Nummer: 22536-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR