7-Fluor-1-tetralon
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
7-Fluoro-3,4-dihydronaphthalen-1(2H)-one
AS33391G1
AS33391G5
ACTIAS33391G1EA
0
CHF
ACTIAS33391G1
ACTIAS33391G5
7-Fluor-1-tetralon
7-Fluor-1-tetralon
Formel:
C₁₀H₉FO MW: 164,18 g/mol |
MDL Number:
MFCD07375433 CAS-Nummer: 2840-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...