6-Brom-2-tetralon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Bromo-3,4-dihydronaphthalen-2(1H)-one

AS30621G1 AS30621G5
ACTIAS30621G1EA 0 CHF
ACTIAS30621G1 ACTIAS30621G5
6-Brom-2-tetralon
6-Brom-2-tetralon
Formel: C₁₀H₉BrO
MW: 225,08 g/mol
MDL Number: MFCD00239388
CAS-Nummer: 4133-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR