2-Fluor-3-nitrobenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS31242G1 AS31242G5
ACTIAS31242G1EA 0 CHF
ACTIAS31242G1 ACTIAS31242G5
2-Fluor-3-nitrobenzoesäure
2-Fluoro-3-nitrobenzoic acid
Formel: C₇H₄FNO₄
MW: 185.11 g/mol
MDL Number: MFCD06797825
CAS-Nummer: 317-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR