(S)-(-)-3-Chlor-1-phenyl-1-propanol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (S)-3-Chloro-1-phenylpropan-1-ol

AS35210G1 AS35210G10 AS35210G5
ACTIAS35210G1EA 0 CHF
ACTIAS35210G1 ACTIAS35210G10 ACTIAS35210G5
(S)-(-)-3-Chlor-1-phenyl-1-propanol
(S)-(-)-3-Chlor-1-phenyl-1-propanol
Formel: C₉H₁₁ClO
MW: 170,64 g/mol
MDL Number: MFCD00013309
CAS-Nummer: 100306-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR