5-Chlor-2-(trifluormethyl)benzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS32512G1 AS32512G5
ACTIAS32512G1EA 0 CHF
ACTIAS32512G1 ACTIAS32512G5
5-Chlor-2-(trifluormethyl)benzaldehyd
5-Chlor-2-(trifluormethyl)benzaldehyd
Formel: C₈H₄ClF₃O
MDL Number: MFCD03412204
CAS-Nummer: 90381-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR