4-Butoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-Butoxyphenyl)boronic acid, 4-n-Butoxyphenylboronic acid

AS50039G1 AS50039G5
ACTIAS50039G1EA 0 CHF
ACTIAS50039G1 ACTIAS50039G5
4-Butoxyphenylboronsäure
4-Butoxyphenylboronsäure
Formel: C₁₀H₁₅BO₃
MW: 194,04 g/mol
MDL Number: MFCD03427054
CAS-Nummer: 105365-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR