(2-Bromphenyl)aceton
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
1-(2-Bromophenyl)propan-2-one
AS32975G1
AS32975G5
ACTIAS32975G1EA
0
CHF
ACTIAS32975G1
ACTIAS32975G5
(2-Bromphenyl)aceton
(2-Bromphenyl)aceton
Formel:
C₉H₉BrO MW: 213,07 g/mol |
MDL Number:
MFCD03410431 CAS-Nummer: 21906-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...