2-Brom-1,3,4-thiadiazol
Lieferant: ACTIVATE SCIENTIFIC
AS33493G1
AS33493G5
ACTIAS33493G1EA
0
CHF
ACTIAS33493G1
ACTIAS33493G5
2-Brom-1,3,4-thiadiazol
2-Bromo-1,3,4-thiadiazole
Formel:
C₂HBrN₂S |
MDL Number:
MFCD04115375 CAS-Nummer: 61929-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...