2-Benzoxazolinon
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
benzo[d]oxazol-2(3H)-one
AS33854G10
AS33854G100
ACTIAS33854G10EA
0
CHF
ACTIAS33854G10
ACTIAS33854G100
2-Benzoxazolinon
2-Benzoxazolinon
Formel:
C₇H₅NO₂ |
MDL Number:
MFCD00005716 CAS-Nummer: 59-49-4 EINECS: 200-430-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...