1,2-Phenylendiessigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: o-Phenylenediacetic acid, 2,2'-(1,2-Phenylene)diacetic acid

AS23895G100 AS23895G25
ACTIAS23895G100EA 270 CHF
ACTIAS23895G100 ACTIAS23895G25
1,2-Phenylendiessigsäure
1,2-Phenylendiessigsäure
Formel: C₁₀H₁₀O₄
MW: 194,19 g/mol
MDL Number: MFCD00004329
CAS-Nummer: 7500-53-0
EINECS: 231-355-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR