Procain
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-(Diethylamino)ethyl 4-Aminobenzoate, 4-Aminobenzoic acid 2-(Diethylamino)ethyl ester, 2-(Diethylamino)ethyl 4-aminobenzoate
AS34421G10
AS34421G100
ACTIAS34421G10EA
0
CHF
ACTIAS34421G10
ACTIAS34421G100
Procain
Procain
Formel:
C₁₃H₂₀N₂O₂ MW: 236,31 g/mol |
MDL Number:
MFCD00007893 CAS-Nummer: 59-46-1 EINECS: 200-426-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...