5-Brom-2-methoxypyrimidin

Lieferant: ACTIVATE SCIENTIFIC

AS50115G100 AS50115G5
ACTIAS50115G100EA 0 CHF
ACTIAS50115G100 ACTIAS50115G5
5-Brom-2-methoxypyrimidin
5-Bromo-2-methoxypyrimidine
Formel: C₅H₅BrN₂O
MW: 189,01 g/mol
MDL Number: MFCD04973351
CAS-Nummer: 14001-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR