1-Methyl-1H-pyrazol-5-boronsäurepinakolester
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 4,4,5,5-Tetramethyl-2-(1-methylpyrazol-5-yl)-1,3,2-dioxaborolane, 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Methylpyrazole-5-boronic acid Pinacol ester
AS32488G1
AS32488G5
ACTIAS32488G1EA
0
CHF
ACTIAS32488G1
ACTIAS32488G5
1-Methyl-1H-pyrazol-5-boronsäurepinakolester
1-Methyl-1H-pyrazol-5-boronsäurepinakolester
Formel:
C₁₀H₁₇BN₂O₂ MW: 208,07 g/mol |
MDL Number:
MFCD05861380 CAS-Nummer: 847818-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...