3-Amino-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole

AS7065G100 AS7065G5
ACTIAS7065G100EA 0 CHF
ACTIAS7065G100 ACTIAS7065G5
3-Amino-1H-pyrazol
3-Amino-1H-pyrazole
Formel: C₃H₅N₃
MW: 83,09 g/mol
MDL Number: MFCD00005236
CAS-Nummer: 1820-80-0
EINECS: 217-338-1
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR